Alpha-Amylase, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219123910
0219123925
IC19123910EA
212.92
USD
IC19123910
IC19123925
Alpha-Amylase, MP Biomedicals
Enzymes
One unit catalyzes the liberation of 1 mmole reducing group (calculated as maltose per minute at pH 6.9 at 20 °C).
- Origin: Product is of animal origin
One unit catalyzes the liberation of 1 mmole reducing group (calculated as maltose per minute at pH 6.9 at 20 °C).