D-(+)-Altrose, white powder
Supplier: 0000012207
D-(+)-Altrose is used as a substrate to identify, differentiate and characterize aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae.
Soluble in water (50 mg/mL - clear, colorless solution); practically insoluble in alcohol.
D-(+)-Altrose is used as a substrate to identify, differentiate and characterize aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae.
Caution: Research Use Only (RUO).
Formula:
C₆H₁₂O₆ |
MDL Number:
MFCD00075620 CAS Number: 1990-29-0 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...