Coomassie® brilliant blue R-250, purple powder C.I. 42660
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Brillant blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-benzenemethanaminium inner salt, sodium salt (1:1)
, Brilliant indocyanine 6B
, Brilliant Blue R
, Serva Blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1)
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt
, Coomassie Blue R 250
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt
, Acid Blue 83
, Brilliant Cyanine 6B
Coomassie Blue R-250 is a triphenylmethane dye used extensively in SDS-PAGE for the analysis of proteins.
Brilliant Blue R has been used for many years for staining proteins on polyacrylamide electrophoresis gels. Improvement of sensitivity and background have been accomplished using Brilliant Blue dyes in a colloidal form. The Brilliant Blue R-Protein complex has an absorption maximum between 560 and 575 nm. (Scanning is usually done at 570 nm.) The staining intensity of proteins may be related to the basicity of the protein involved. Other applications for brilliant Blue R include use in detection of lipids in thin layer chromatography, determination of the CMC of detergents, and detection of proteins in solution, as well as on cellulose acetate plates. Coomassie Blue R-250 is additionally used in the Bradford Assay to quantify the concentration of protein in a solution, such as cell lysates. A fast acting, sensitive dye which can be used on SDS gels, IEF gels, and standard gels.
The dye molecule binds to proteins to form a protein-dye complex. This complex stabilizes the negatively charged anionic form of the dye producing the blue color, even under acidic conditions.
Dark blue-black solution(10 mg/ml of water). Soluble in water (70 mg/ml); ethanol (10 mg/ml); 20 mg/ml in methyl cellosolve. Prepare in distilled, deionised water: 50% (v/v) methanol, 0,05% (w/v) Coomassie Brilliant Blue R-250, 10% acetic acid, 40% Water. Dissolve the Coomassie Brilliant Blue R in methanol before adding acetic acid and water (filtration is not generally needed). Solution can be stored for 6 months. If precipitate is observed following prolonged storage, filter to obtain a homogeneous solution.
Formula:
C₄₅H₄₄N₃NaO₇S₂ MW: 825.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041762 CAS Number: 6104-59-2 |
Specification Test Results
Identity Test | Passes |
l max (ethanol) | 585 ± 5 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...