Methacholine chloride ≥98%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Mecholyl Chloride, O-Acetyl-beta-methylcholine Chloride, Acetyl-β-methylcholine chloride
Methacholine chloride is a cholinergic agonist and parasympathomimetic bronchoconstrictor. A metabolically stable analog of acetylcholine. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.
Methacholine is primarily used to diagnose bronchial hyperreactivity, which is the hallmark of asthma and also occurs in chronic obstructive pulmonary disease. Acetyl-β-methylcholine (methacholine), an analog of acetylcholine, is use as a receptor agonist to study processes such as airway hyperresponsiveness associated with asthma.
Metabolically stable analog of acetylcholine; muscarinic agonist. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.
- Presentation: White Powder
- Soluble in water (100 mg/mL-clear, colorless solution), alcohol, chloroform; Insoluble in ether.
- -20°C, desiccate
Formula:
C₈H₁₈ClNO₂ Melting Pt: 164…174 °C Storage Temperature: Freezer |
MDL Number:
MFCD00011817 CAS Number: 62-51-1 EINECS: 200-537-2 |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...