Hypotaurine, white prills

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2-Aminoethane-1-sulphinic acid

0215971180 0215971183
IC15971180EA 71.64 USD
IC15971180 IC15971183
Hypotaurine, white prills
Hypotaurine

Hypotaurine is a Non-specific, endogenous inhibitory glycine receptor agonist.


An antioxidant that binds hydroxyl radicals. Endogenous inhibitory neuromodulator that is blocked by strychnine and picrotoxin. Inhibits the Na2+-dependent transport of GABA and β-alanine. It is a degradation product of cysteine and a precursor of taurine.


Soluble in water (100 mM - colorless to faint yellow solution, 100 mg/mL).


Antioxidant that binds hydroxyl radicals. Endogenous inhibitory neuromodulator that is blocked by strychnine and picrotoxin. Inhibits the Na2+-dependent transport of GABA and β-alanine. It is a degradation product of cysteine and a precursor of taurine.

Formula: C₂H₇NO₂S
MW: 109.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038197
CAS Number: 300-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR