Spectinomycin sulphate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215899302 0215899301 0215899305
IC15899302EA 99.58 USD
IC15899302 IC0215899301 IC0215899305
Spectinomycin sulphate
Spectinomycin sulphate

Spectinomycin sulfate is a broad spectrum antibiotic that inhibits protein synthesis (elongation) by binding to the bacterial 30S ribosomal subunit and interfering with peptidyl tRNA translocation.


  • White powder


Soluble in DMSO, ethanol and water; practically insoluble in pyridine, chloroform,ethyl acetate, cyclohexone, benzene, acetone, DMF,dioxane and acetonitrile.

Formula: C₁₄H₂₄N₂O₇·H₂SO₄
MW: 430.43 g/mol
Melting Pt: 189…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270185
CAS Number: 23312-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR