Cyclopiazonic acid 98, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215889505 0215889510
IC15889505EA 139.16 USD
IC15889505 IC15889510
Cyclopiazonic acid 98, powder
Cyclopiazonic acid

Cyclopiazonic acid on Langendorff-perfused murine hearts displays anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis.


Inhibits the sarcoplasmic reticulum Ca2+-pump.


Soluble in chloroform (10 mg/mL) or methanol (10 mg/mL - clear, bright yellow solution).


Cyclopiazonic acid on Langendorff-perfused murine hearts displays anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis.

Formula: C₂₀H₂₀N₂O₃
MW: 336.391 g/mol
MDL Number: MFCD00167445
CAS Number: 18172-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR