(+/-)-Thalidomide ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-(2,6-Dioxopiperidin-3-yl)isoindoline-1,3-dione, N-(2,6-Dioxo-3-piperidinyl)phthalimide

0215875350 0215875383
IC15875350EA 66.36 USD
IC15875350 IC15875383
(+/-)-Thalidomide ≥99%, powder
(+/-)-Thalidomide

(±)-Thalidomide selectively inhibits biosynthesis of tumor necrosis factor α (TNF-α); inhibitor of angiogenesis; immunosuppressive; sedative; teratogen.


Soluble in DMSO (>20 mg/ml).


(±)-Thalidomide selectively inhibits biosynthesis of tumor necrosis factor α (TNF-α); inhibitor of angiogenesis; immunosuppressive; sedative; teratogen.

Formula: C₁₃H₁₀N₂O₄
MDL Number: MFCD00153873
CAS Number: 50-35-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR