Zincon sodium salt, brown powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215831701 0215831705
IC15831701EA 112.04 USD
IC15831701 IC15831705
Zincon sodium salt, brown powder
Zincon sodium salt

Zincon is a colorimetric indicator for Zn, Cu and Hg. Reagent for the spectrophotometric determination of zinc. Simultaneous determination of zinc and copper. Used in automated flow injection analysis of zinc. As a metal indicator for EDTA titration of Zn, or for the back titration of Ca in the presence of Mg with glycoletherdiaminetertraacetic acid (GEDTA or EGTA).


Reacts with Zn (pH 8,5 to 9,5) and Cu (pH 5,0 to 9,5) to form blue chelates. Also color reaction occurs with Co, Ni and some other polyvalent cations, Beer's law is observed in the range of 0,1 to 2,4 μg Cu/ml in water at 600 mu.


Slightly soluble in water and alcohol. Insoluble in most organic solvents. Soluble in aqueous alkali resulting in a red solution.


Zincon is a colorimetric indicator for Zn, Cu and Hg. Reagent for the spectrophotometric determination of zinc. Simultaneous determination of zinc and copper. Used in automated flow injection analysis of zinc. As a metal indicator for EDTA titration of Zn, or for the back titration of Ca in the presence of Mg with glycoletherdiaminetertraacetic acid (GEDTA or EGTA).

Formula: C₂₀H₁₅N₄NaO₆S
Melting Pt: 250…260 °C (dec., lit.)
MDL Number: MFCD00064385
CAS Number: 62625-22-3

Order Now

Specification Test Results

Identity Test Passes
Storage Store at Room Temperature (15-30 °C).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR