n-Undecane ≥99%, clear, colorless liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Undecane
Undecane may be used as an internal standard in gas chromatography when working with other hydrocarbons. Since the boiling point of undecane (196 °C) is well known, it may be used as a comparison for retention times in a gas chromatograph for molecules whose structure has been freshly elucidated.
Japan Fire Code:4.2
Undecane may be used as an internal standard in gas chromatography when working with other hydrocarbons. Since the boiling point of undecane (196 °C) is well known, it may be used as a comparison for retention times in a gas chromatograph for molecules whose structure has been freshly elucidated.
Formula:
H₃C(CH₂)₉CH₃ MW: 156.31 g/mol |
MDL Number:
MFCD00008959 CAS Number: 1120-21-4 UN: 2330 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...