Iron(II) chloride tetrahydrate, crystalline
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate
0215804683
0215804691
IC15804683EA
140.69
USD
IC15804683
IC15804691
Iron(II) chloride tetrahydrate, crystalline
Iron (II) chloride tetrahydrate
Soluble in water, alcohol, acetone.
Ferrous chloride is used in metallurgy as an electrolytic agent and as a mordant in dyeing. It has been used in the synthesis of pentasubstituted acylferrocenes. In biological research, ferrous chloride is used as a source of Fe2+ ion.
Formula:
FeCl₂·4H₂O MW: 198.81 g/mol Melting Pt: 677 |
MDL Number:
MFCD00149709 CAS Number: 13478-10-9 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...