Eosin Y, orange powder C.I. 45380:2
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
Eosin Y is a counterstain for hematoxylin.
It is used in combination with Azure II to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; bond's modification of mann's eosin-methyl blue for negri bodies in smears; harris stain for negri bodies in smears; mann's method for staining negri bodies in sections; gridley's stain for endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of endamoeba in sections.
Soluble in water (0,8 mg/ml); ethanol (10 mg/ml); ethylene glycol monomethyl ether (methyl-cellosolve; EGME) (40 mg/ml).
Eosin Y is a counterstain for hematoxylin. It is used in combination with Azure II to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; Bond's modification of Mann's Eosin-Methyl Blue for Negri bodies in smears; Harris stain for Negri bodies in smears; Mann's method for staining Negri bodies in sections; Gridley's stain for Endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of Endamoeba in sections.
Formula:
C₂₀H₈Br₄O₅ |
MDL Number:
MFCD00036189 CAS Number: 15086-94-9 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...