Sesame oil, colourless oil

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215662183 0215662191
IC15662183EA 76.53 USD
IC15662183 IC15662191
Sesame oil, colourless oil
Sesame oil

Sesame oil is a potent antioxidant, which can inhibit DNA damage and lipid peroxidation by 4-Nitroquinoline N-oxide in rats.


Studies indicate that Sesame oil can increase wound healing in SKH1 mice when added together with ozone. It is known that Sesame oil is composed of triacylglycerols encapsulated by a monolayer of phospholipids which is then covered by the proteins oleosin, caleosin and steroleosin. Furthermore, this compound is a great research tool for reversing and preventing oxidative damage on cells and tissues.
Sesame oil used in research applications as delivery vehicle for fat-soluble compounds including steroidal hormones and toxins. Sesame oil has antioxidant properties and was shown to protect against lipopolysaccharide (LPS)-stimulated oxidative stress in animal models.


Soluble in chloroform, ether, petroleum ether, carbon disulphide; slightly soluble in alcohol; insoluble in water.

MDL Number: MFCD00132243
CAS Number: 8008-74-0

Order Now

Specification Test Results

Synonyms Benne oil; Teel oil; Gingilli oil
Form Clear, Colorless Oil
Storage Store at room Temperature(15-30 °C).
Refractive Index 1.4650-1.4690 at 40 °C (Lit.)
Source Sesamum indicum seeds


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR