Propylene carbonate, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Carbonic acid propylene glycol ester
, 4-Methyl-1,3-dioxolan-2-one
, Propylene glycol carbonate
0215641594
0215641590
IC15641590EA
108.48
USD
IC15641590
IC15641594
Propylene carbonate, liquid
Propylene carbonate
Soluble in water.
Formula:
C₄H₆O₃ MW: 102.09 g/mol |
MDL Number:
MFCD00005385 CAS Number: 108-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...