Nisin

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215583905 0215583925
IC15583905EA 238.17 USD
IC15583905 IC15583925
Nisin
Nisin

Nisin is a polypeptide (34 residues) antibiotic structurally similar to subtilin but containing no tryptophan; a member of the lantibiotic family. Reduces the thermal resistance of bacterial spores in milk-based beverages.The C-terminus of nisin is responsible for the initial interaction of nisin, binding to the target membrane. Active against Gram-positive bacteria at the cytoplasmic membrane.


Antibiotic with bactericidal action. Binds to the lipid A portion of bacterial lipopolysaccharides. Induces pore formation in the membranes of cortex cells from excised sorghum roots. Mode of Action: Binds to and interferes with the permeability of the cytoplasmic membrane. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria (requires chelator such as EDTA).


Soluble in dilute acids such as 0.02 N HCl (118 mg/mL). Any undissolved material is the dairy solids and should not be considered a problem. Stable to boiling in acid solutions.


Nisin is a polypeptide (34 residues) antibiotic structurally similar to subtilin but containing no tryptophan; a member of the lantibiotic family. Reduces the thermal resistance of bacterial spores in milk-based beverages.The C-terminus of nisin is responsible for the initial interaction of nisin, binding to the target membrane. Active against Gram-positive bacteria at the cytoplasmic membrane.

Formula: C₁₄₃H₂₃₀N₄₂O₃₇S₇
MW: 3354.06 g/mol
MDL Number: MFCD00131724
CAS Number: 1414-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e67aca2ac5b8ce32bcf086ab349dfcb13e0fd68&vid=4e67aca2ac5b8ce32bcf086ab349dfcb13e0fd68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Ambicin N
Concentration Nisin content: 2.5% .Remainder is NaCl and milk solids (denatured).
Form Tan Powder
Storage Store at Room Temperature (15-30 °C).
pH 3.31 (10% aq suspension) (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR