1-Naphtholbenzeine, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
alpha-Naphtholbenzein
, α-Naphtholbenzein
, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one
, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol
, α-Naphtholbenzeine
, p-Naphtholbenzeine
, 1-Naphtholbenzein
, Bis(4-hydroxy-1-naphthyl)phenylmethanol
, p-Naphtholbenzein
0215578505
0215578525
IC15578525EA
208.71
USD
IC15578525
IC15578505
1-Naphtholbenzeine, Powder
1-Naphtholbenzeine
Soluble in ethanol, acetic acid.
Visual Transition: orange to blue (pH 8 to pH10)
Formula:
C₂₇H₁₈O₂ MW: 374.44 g/mol |
MDL Number:
MFCD00078492 CAS Number: 145-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...