Methyl ethyl ketone, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

0215554891
IC15554891EA 106.26 USD
IC15554891
Methyl ethyl ketone, liquid
Methyl ethyl ketone

Soluble in alcohol, ether and most fixed oils. Soluble in water.

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR