Melanin
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Melanin is a natural substance that gives color (pigment) to hair, skin, and the iris of the eye. It is produced by cells in the skin called melanocytes. Melanin also helps protect the skin from the sun.Melanin is an extremely stable molecule, and highly resistant to the digestion by most acids and bases.
Melanin is used to prepare inhibitor in Real Time PCR Analysis.
Prepared by persulfate oxidation of tyrosine.
Soluble in dilute NH3
Melanin is a natural substance that gives color (pigment) to hair, skin, and the iris of the eye. It is produced by cells in the skin called melanocytes. Melanin also helps protect the skin from the sun.Melanin is an extremely stable molecule, and highly resistant to the digestion by most acids and bases.
Formula:
C₁₈H₁₀N₂O₄ |
MDL Number:
MFCD00131581 CAS Number: 8049-97-6 |
Specification Test Results
Form | Dark Gray Powder |
Storage | Store at - 0 °C |
Source | Synthetic |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...