Manganese(II) sulfate monohydrate, pale pink powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

0215533690 0215533691
IC15533690EA 91.09 USD
IC15533690 IC15533691
Manganese(II) sulfate monohydrate, pale pink powder
Manganese (II) sulphate monohydrate

Magnesium sulfate is used as a colorant in dyes and red glazes for porcelain. As a trace essential element it is a nutritional factor for all animals; often a supplement in feed. It is used in biological assays to study the effects of manganese on the central nervous system, and to assess exposure limits.


Soluble in water (77 mg/mL), yielding a clear, light pink solution.(Lit.)


Magnesium has a variety of biological roles in enzymology, cell membrane and wall structural integrity, muscle cell physiology, and nucleic acid structure. Magnesium is an essential co-factor in many enzymes, including deoxyribonuclease (DNase), the restriction enzymes EcoR I and EcoR V, and Ribonuclease H.Magnesium also stabilizes polymeric nucleic acids such as transfer RNA and ribozymes.

Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
MDL Number: MFCD00149159
CAS Number: 10034-96-5
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR