Acetic anhydride, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Acetyl oxideAcetyl acetate

0215468091 0215468090
IC15468090EA 86.98 USD
IC15468090 IC15468091
Acetic anhydride, liquid
Acetic anhydride

Miscible in chloroform and ether. In alcohol, it will form ethyl acetate. In water (0.1 mL/mL, v/v), this product will slowly form acetic acid.


expl. lim. 10.3 % (Lit.); Molarity: 10.6 M (calculated from density and molecular weight)


Acetic anhydride is a reagent that is used in the manufacturing of acetyl compounds and cellulose acetates. It is also utilized as a solvent in the examination of wool fat, glycerol, fatty and volatile oils, and resins. In the synthesis of organic compounds, acetic anhydride is used in various reactions, including the cleavage of dialkyl ethers, the formation of anhydrides from carboxylic acids, the addition of methylthiomethyl groups to phenol rings, and the addition of acyl and acyloxy groups across double bonds. Acetic anhydride has been used to hyperacetylate histones to investigate transcriptional effects of the NAD-dependent histone deacetylase Sir2 in Drosophila on a complex of the hyperacetylated histones with DNA. Acetic anhydride is utilized in an assay of a fluorescently labeled peptide catalyst library for application to the resolution of secondary alcohols.

Formula: (CH₃CO)₂O
MW: 102.09 g/mol
MDL Number: MFCD00008705
CAS Number: 108-24-7
UN: 1715
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/99/75/36419975.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR