Lactulose ~98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215393205 0215393225 0215393280
IC15393205EA 95.23 USD
IC15393205 IC15393225 IC15393280
Lactulose ~98%, white powder
Lactulose

Lactulose is a synthetic, non-digestible sugar used in the treatment of chronic constipation and hepatic encephalopathy, a complication of liver disease. It is a disaccharide (double-sugar) formed from one molecule each of the simple sugars (monosaccharides) fructose and galactose.


Lactulose is used as a test of small intestine bacterial overgrowth (SIBO). Lactulose is used in the treatment of chronic constipation. It is useful in treating the hyperammonemia caused by hepatic encephalopathy, because it helps "draw out" ammonia from the body. Lactulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health.


Soluble in 5% of water (colorless)


Lactulose is a synthetic, non-digestible sugar used in the treatment of chronic constipation and hepatic encephalopathy, a complication of liver disease. It is a disaccharide (double-sugar) formed from one molecule each of the simple sugars (monosaccharides) fructose and galactose.

Formula: C₁₂H₂₂O₁₁
MW: 342.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151469
CAS Number: 4618-18-2

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
FTIR Conforms to Reference
Moisture Content 3,0%
Optical Rotation -45 to -52° (c=5, water, 24 hrs)
pH (1% Aqueous solution) 5,5 - 6,5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR