Methylene Green zinc chloride double salt, powder_dark green C.I. 52020
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Basic Green 5, C.I. 52020, 3,7-Bis(Dimethylamino)-4-nitro-Phenothiazin-5-Ium chloride, Methylene Green complex with half ZnCl2
0215264405
0215264480
IC15264405EA
53.52
USD
IC15264405
IC15264480
Methylene Green zinc chloride double salt, powder_dark green C.I. 52020
Methylene Green zinc chloride double salt
Dark Green almost Black Fine Powder
Soluble in water (10 mg/ ml-opaque, deep blue solution) and alcohol.
Formula:
C₁₆H₁₇ClN₄O₂S·0.5ZnCl₂ MW: 433.00 g/mol |
MDL Number:
MFCD00151524 CAS Number: 6722-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...