Trichloroacetic acid (TCA) ≥99.0%, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
TCA, Trichloroethanoic acid, Aceto-Caustin, TCA
Trichloroacetic acid is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation. Used as a decalcifier and fixative in microscopy. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.
Stability: Long term storage of aqueous solutions of less than 30% strength is not recommended. Decomposition products are chloroform, hydrochloric acid, carbon dioxide and carbon monoxide. Chloride ≤0.001% ; Nitrate ≤0.002%; Phosphate ≤5 ppm; Sulfate ≤0.02%
- ACS Grade
- Vapor Density: 5.6 (air = 1)(Lit.)
- Vapor Pressure: 1.2 hPa at 50 °C(Lit.)
- Refractive Index: n20/D 1.62(Lit.)
- pH of a 0.1 M aqueous solution is 1.2(Lit.)
- Soluble in water; very soluble in ethanol or ether. Decomposed by heating with caustic alkalies into chloroform and alkali carbonate.
- Storage temperature: Store at Room Temperature (15-30 °C). Store Desiccated.
Stability: Long term storage of aqueous solutions of less than 30% strength is not recommended. Decomposition products are chloroform, hydrochloric acid, carbon dioxide and carbon monoxide. Chloride ≤0.001% ; Nitrate ≤0.002%; Phosphate ≤5 ppm; Sulfate ≤0.02%
Formula:
Cl₃CCOOH Boiling Pt: 197 °C (1013 hPa) (lit.) Melting Pt: ∼56 °C (lit.) Density: 1.61 g/cm³ (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 UN: 1839 ADR: 8,II |
|
Specification Test Results
Purity | ≥99.0% |
Residue after Ignition | ≤0.03% |
Chloride | ≤0.002% |
Nitrate | ≤0.002% |
Phosphate | ≤5ppm |
Sulfate | ≤0.02% |
Heavy Metals | ≤0.002% |
Iron | ≤0.001% |
Substance Darkened by Sulfuric Acid | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...