Potassium thiocyanate ≥98.6%, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Potassium rhodanide
0215256680
0215256690
IC15256680EA
107.75
USD
IC15256680
IC15256690
Potassium thiocyanate ≥98.6%, powder ACS
Potassium thiocyanate
Soluble in water (100 mg/mL), yielding a clear, colorless solution. The aqueous dissolution of KSCN is an endothermic process and gives solutions of neutral pH. KSCN is also soluble in alcohol (83 mg/mL) and acetone (2 g/mL).
Formula:
KSCN MW: 97.18 g/mol Boiling Pt: 500 Melting Pt: 175 (lit.) |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 Merck Index: 12,07866 |
Specification Test Results
Assay | 98.64% |
pH (5% soln) | 6.84 |
Insoluble in Water | 0.00% |
Chloride (Cl) |
|
Sulfate (SO4) |
|
Ammonium (NH4) |
|
Heavy Metals (as Pb) |
|
Iron (Fe) |
|
Sodium (Na) | 0.00% |
Iodine Consuming Substances | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...