Hydroxy naphthol blue disodium salt ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulfo-3-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulfonate, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

0215252910 0215252980
IC15252910EA 57.76 USD
IC15252910 IC15252980
Hydroxy naphthol blue disodium salt ACS
Hydroxy naphthol blue disodium salt

Hydroxynaphthol blue, disodium salt is used for determining the endpoint in metal complexation titrations.


Soluble in water (clear, red solution).


Hydroxynaphthol blue, disodium salt is used for determining the endpoint in metal complexation titrations.

Formula: C₂₀H₁₂N₂O₁₁S₃·2Na
MDL Number: MFCD00004075
CAS Number: 165660-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR