D-(+)-Glucose, anhydrous, white powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Corn sugar
, D-Glucose
, D-(+)-Dextrose
, Glucolin
, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
, Glucose
, Dextrose
0215252794
0215252790
IC15252794EA
339.13
USD
IC15252794
IC15252790
D-(+)-Glucose, anhydrous, white powder ACS
D(+)-Glucose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 150…152 °C (lit.) |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 |
Specification Test Results
Identity Test | Passes |
Specific Rotation | 52.5 to 53.0° |
Insoluble Matter |
|
Loss on Drying |
|
Residue on Ignition |
|
Titratable Acid |
|
Chloride |
|
Sulfate (SO₄) and Sulfite |
|
Starch | Passes |
Heavy Metals (PB) | ≤5 ppm, |
Iron | ≤5 ppm, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...