Ammonium iron (II) sulfate hexahydrate 98.5-101.5%, crystalline ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Ferrous Ammonium sulphate Hexahydrate
, Ferrous ammonium sulfate hexahydrate
, Ammonium iron(Ⅱ) sulfate hexahydrate
, Ammonium ferrous sulphate hexahydrate
, Ferrous Ammonium sulfate Hexahydrate
, di-Ammonium iron (II) sulphate hexahydrate
, Ammonium ferrous sulfate hexahydrate
, Ferrous ammonium sulphate hexahydrate
, Ammonium iron(II) sulphate hydrate
, di-Ammonium iron (II) sulfate hexahydrate
, Ammonium iron(Ⅱ) sulphate hexahydrate
, Ammonium iron(II) sulfate hydrate
Ferrous ammonium sulfate is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It is used for titration purposes.
Soluble in water; practically insoluble in alcohol.
Caution: Research Use Only (RUO).
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 |
Specification Test Results
Identity Test | Passes |
Assay | 98,5 - 101,5% |
Insoluble Matter | ≤0,01% |
Phosphate (PO4) |
|
Copper (Cu) |
|
Manganese (Mn) |
|
Zinc (Zn) |
|
Ferric Ion (Fe3+) |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...