EDTA disodium salt dihydrate ≥99%, white crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
Ethylenediamine Tetraacetic Acid is a chelator of divalent cations. It also inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Soluble in water : 0.1 M, clear, colorless aqueous solution.
Ethylenediamine Tetraacetic Acid is a chelator of divalent cations. It also inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Caution: Research Use Only (RUO).
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 |
|
Specification Test Results
Purity | ≥99% |
pH (5% aq soln) | 4 - 6 |
Insoluble Matter | ≤0,05% |
Nitrilotriacetic Acid | ≤0,1% |
Heavy Metals (as Pb ) | ≤0,005% |
Iron (Fe) | ≤0,01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...