Ammonium iodide, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215249680
0215249690
IC15249680EA
127.38
USD
IC15249680
IC15249690
Ammonium iodide, powder ACS
Ammonium iodide
Chloride & Bromide (as Cl) ≤0.005%; Phosphate ≤0.001%; Sulfate ≤0.05%; Barium ≤0.002%; Iron ≤5 ppm
Formula:
NH₄I MW: 144.94 g/mol Melting Pt: 551 |
MDL Number:
MFCD00011424 CAS Number: 12027-06-4 Merck Index: 13,00531 |
Specification Test Results
Form | Powder |
Storage | Store at Room Temperature (15-30 °C), desiccate, protect from light. |
Assay | ≥99.0% |
Heavy Metals | ≤0.001% |
Insoluble Matter | ≤0.005% |
Residue on Ignition | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...