Vanillin ≥99%, off-white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
4-Hydroxy-3-methoxybenzaldehyde
0215218880
0215218890
0215218891
IC15218880EA
96.38
USD
IC15218880
IC15218890
IC15218891
Vanillin ≥99%, off-white powder
Vanillin
Solule in alcohol(50 mg/mL (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Solule in alcohol(50 mg/mL (5%); Ethanol), chloroform and ether; slightly soluble in water and glycol.
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 285 Melting Pt: 82 Flash Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006942 CAS Number: 121-33-5 EINECS: 204-465-2 Merck Index: 13,09998 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...