Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0%, Reagent Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Trizma(R) base
, TRIS base
, THAM
, 2-Amino-2-(hydroxymethyl)-1,3-propanediol
, Tris(hydroxymethyl)methylamine
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris)
, Tromethamine
02152176.1
02152176.5
0215217601
0215217610
0215217605
IC15217601EA
120.33
USD
IC15217601
IC152176.1
IC15217610
IC15217605
IC152176.5
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0%, Reagent Grade
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
40% in water.
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 |
Specification Test Results
Synonyms | Trometamol; Tromethamine; Tris-[hydroxymethyl]aminomethane; 2-Amino-2-(hydroxymethyl)-1,3-propanediol |
Form | White Crystalline Powder |
Storage | Store at Room Temperature (15 - 30 °C), desiccate. |
pH | 10 - 11.5 (5% aq soln) |
pka | 8.1 (25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...