Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%, colorless solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

02152070.5 0215207001 0215207005
IC152070.5EA 79.24 USD
IC152070.5 IC15207001 IC15207005
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%, colorless solid
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)

Spermine is a polyamine involved in cellular metabolism found in all eukaryotic cells. Formed from spermidine through the addition of aminopropyl group to spermidine by spermine synthase. It is found as a polycation at physiological pH.


Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).


Soluble in water (1% w/v), lower alcohol, chloroform; practically insoluble in ether, benzene, petroleum ether.


Caution: Research Use Only (RUO). Desiccate, store under nitrogen.

Formula: C₁₀H₂₆N₄
MW: 202.35 g/mol
Boiling Pt: 141…142 °C (0.5 mmHg)
Melting Pt: 28…30 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00008215
CAS Number: 71-44-3

Order Now

Specification Test Results

Identity Test Passes
Purity ≥97%
FTIR Conforms to Standard
pH (0.5% aq soln) 11 - 14


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR