(±)-1,2-Propanediol, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Propylene glycol is a clear, colorless and hygroscopic liquid. Propylene glycol contains an asymmetrical carbon atom, so it exists in two stereoisomers.
The freezing point of water is depressed when mixed with propylene glycol due to increased opportunity for hydrogen bonding.
Propylene glycol is a solvent in many pharmaceuticals, including oral, injectable and topical formulations, such as for diazepam and lorazepam that are insoluble in water, use propylene glycol as a solvent in their clinical, injectable forms. It is also used as automotive antifreeze.
Formula:
CH₃CH(OH)CH₂OH Boiling Pt: 187.9 °C (1013 hPa) (lit.) Melting Pt: –60 °C (lit.) Density: 1.04 g/cm³ (20 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 |
|
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...