Ponceau S ≥75% (dye content), dark powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
CI 27195
, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt
, Ponceau
, 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt
, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt
Ponceau S is used as protein stain for electrophoresis.
Readily soluble in water.
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄ MW: 760.56 g/mol Melting Pt: 300 °C (lit.) |
MDL Number:
MFCD00003892 CAS Number: 6226-79-5 |
Specification Test Results
Identity Test | Passes |
Dye Content | ~75% |
UV/VIS: l max (water) | 517 - 523 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...