Polyvinyl alcohol, M.W. ~22000, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215193883 0215193891
IC15193883EA 98.39 USD
IC15193883 IC15193891
Polyvinyl alcohol, M.W. ~22000, powder
Polyvinyl alcohol

Polyvinyl alcohol is the raw material to make polymers. Polyvinyl alcohol is used as a emulsion polymerization aid, as protective colloid, to make polyvinyl acetate dispersions.


Viscosity (4% aq soln) 4 - 6 cps


Caution: Research Use Only (RUO).

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc65f96819721d0e2780d697ccf72ab3863bb5e&vid=5fc65f96819721d0e2780d697ccf72ab3863bb5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Viscosity (4% aq soln) 4 - 6 cps


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR