Diethylenetriaminepentaacetic acid ≥90%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
DTPA
, Idranal V
, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid
, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid
, Diethylenetriaminepentaacetic acid
, N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid)
, Kestranal® Aza Pa
, Titriplex® V
Diethylenetriamine-N,N,N',N",N"-pentaacetic Acid is used as a chelating agent. Diethylenetriaminepentaacetic acid may be used to synthesize chitosan (CH)-gadolinium (Gd) diethylenetriaminepentaacetic acid (DTPA) conjugates.
- Presentation: White Powder
- pH: 2.27 (Lit.)
- Soluble in 1 M hydrochloric acid (50 mg/mL) & water.
- Storage temperature: Store at Room Temperature (15-30 °C)
Formula:
C₁₄H₂₃N₃O₁₀ MW: 393.35 g/mol Melting Pt: 219…220 °C (lit.) Density: 1.770 g/mL (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00004289 CAS Number: 67-43-6 ADR: 9,III |
Specification Test Results
Identity Test | Passes |
Purity | ~90% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...