Glycerine, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol
0215119490
0215119491
IC15119490EA
69.04
USD
IC15119490
IC15119491
Glycerine, liquid
Glycerine
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 290 Melting Pt: 18.6 Flash Pt: 177 |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 Merck Index: 13,04497 |
|
Specification Test Results
Synonyms | 1,2,3-Propanetriol; Glycerin; trehydroxypropane |
Form | Liquid |
Storage | Store at Room Temperature (15-30 °C). |
pH | 5.0 -7.5 (1% aq soln) |
Refractive Index | n20/D 1.474 (Lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...