Congo red, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28

0215071105 0215071180
IC15071105EA 59.63 USD
IC15071105 IC15071180
Congo red, powder
Congo Red

Soluble in water (10 mg/mL).

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
MDL Number: MFCD00004028
CAS Number: 573-58-0
Merck Index: 12,02562

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR