Brilliant cresyl blue, dark green powder C.I. 51010
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
BCB
, Brilliant Blue C
, Brilliant cresyl blue zinc chloride double salt
, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1)
, CI 51010
Brilliant Cresyl Blue is a supravital stain used in hematology for demonstrating and counting reticulocytes and platelets. It is also used as a vital stain for demonstrating the mucous secretions of the marine coelomates.
Soluble in water, 1 N Hydrochloric acid, or 50% ethanol.
Satisfactory for staining reticulocytes in fresh blood using both aqueous and alcoholic stain preparations.
Formula:
C₃₄H₄₂Cl₄N₈O₂Zn MW: 801.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00148901 CAS Number: 81029-05-2 |
Specification Test Results
Identity Test | Passes |
UV/Vis: l max (ethanol) | For Information Only |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...