(1S)-(-)-Borneol, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
(1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
, (-)-Borneol
, L(-)-Borneol
0215051310
0215051380
IC15051380EA
177.77
USD
IC15051380
IC15051310
(1S)-(-)-Borneol, white crystalline powder
(1S)-(-)-Borneol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol |
MDL Number:
MFCD00003759 CAS Number: 464-45-9 UN: 1312 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...