Bismarck brown Y, dark brown powder C.I. 21000

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: C.I. 21000, 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride, Bismarck Brown Y (G), Basic Brown 1, Vesuvine, Phenylene Brown, Leather Brown, Basic Brown G, GX, GXP, Manchester Brown, 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

0215047325 0215047380
IC15047325EA 65.77 USD
IC15047325 IC15047380
Bismarck brown Y, dark brown powder C.I. 21000
Bismarck Brown Y

Bismarck brown Y is a Plasma stain used for staining mucine, cartilage and goblet cells.


Soluble in water, ethanol, acetic acid, nitric acid; slightly soluble in ethanol and celosolve; practically insoluble in acetone, benzene carbon tetrachloride.

Formula: C₁₈H₁₈N8·2HCl
MW: 419.32 g/mol
MDL Number: MFCD00012977
CAS Number: 10114-58-6
Merck Index: 12,01294

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: l max 452 - 457 nm (50% EtOH, few drops of 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR