Tin(II) chloride ∼99%, anhydrous, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Stannous chloride, Tin dichloride
0215015780
0215015783
0215015790
IC15015780EA
111.14
USD
IC15015780
IC15015783
IC15015790
Tin(II) chloride ∼99%, anhydrous, white crystalline powder
Tin (II) chloride
Stannous Chloride is used as a reducing agent, as a reagent in analytical chemistry and as a metal catalyst in organic reactions.
Formula:
SnCl₂ MW: 189.62 g/mol Boiling Pt: 652 °C (lit.) Melting Pt: 246 °C |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 UN: 3260 ADR: 8,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...