Paraformaldehyde, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Polyoxymethylene

02150146.5 0215014601
IC15014601EA 103.5 USD
IC15014601 IC150146.5
Paraformaldehyde, powder
Paraformaldehyde

Soluble in water (hot water - ~37% w/v; slowly soluble in cold water). Aqueous solutions will develop formaldehyde over time. Insoluble in alcohol or ether.

Formula: (CH₂O)n
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1,III
https://us.vwr.com/stibo/web/std.lang.all/00/33/36380033.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR