Chloramine-T (sodium salt) trihydrate, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, N-Chloro-4-toluenesulfonamide sodium salt, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
0215011580
0215011590
76204-104EA
48.12
USD
76204-104
76204-106
Chloramine-T (sodium salt) trihydrate, white crystalline powder
Chloramine T (sodium salt) trihydrate
Soluble in water (100 mg/mL); practically insoluble in benzene, chloroform or ether.
- Presentation : White Crystalline Powder
- pH : 6 - 9(1% aq soln)
- Detection Method : FTIR
- Solubility :
Storage: protect from light, under nitrogen.
Formula:
CH₃C₆H₄SO₂NClNa·3H₂O MW: 281.69 g/mol Melting Pt: 167...170 °C (dec., lit.) Density: 540...680 kg/m³ (bulk density) Flash Pt: 192 °C (closed cup) Storage Temperature: Refrigerator |
MDL Number:
MFCD00149066 CAS Number: 7080-50-4 UN: 3263 ADR: 8,III |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH (5% aq soln) | 8 - 10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...