Ammonium chloride, powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Ammonium muriate
, Salmiac
0215010980
IC15010980EA
46.57
USD
IC15010980
Ammonium chloride, powder
Ammonium chloride
Soluble in water (100 mg/mL- clear, colorless solution). The presence of hydrochloric acid or sodium chloride decrease water solubility. Also soluble in methanol and ethanol.
Formula:
NH₄Cl MW: 53.49 g/mol |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...