N-Octyl-β-D-glucopyranoside ≥98%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
n-Octyl-β-D-Glucopyranoside is a non-ionic detergent for membrane protein.
n-Octyl-glucoside is a non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids. n-Octyl-glucoside has been used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers. n-Octyl-glucoside has been used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.
Soluble in water (10 mg/mL-clear colorless solution). 20-25 mM Detergents with high CMC values are generally easy to remove by dilution; detergents with low CMC values are advantageous for separations on the basis of molecular weight. As a general rule, detergents should be used at their CMC and at a detergent-to-protein weight ratio of approximately ten.
Formula:
C₁₄H₂₈O₆ MW: 292.37 g/mol Melting Pt: 89…100 °C (lit.) Storage Temperature: Freezer |
MDL Number:
MFCD00063288 CAS Number: 29836-26-8 |
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Optical Rotation (c=5, water) | -31 ± 3° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...