Urea ≥99%, white powder, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

0210569501 0210569505
IC10569501EA 60.52 USD
IC10569501 IC10569505
Urea ≥99%, white powder, Ultrapure
Urea

Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex.


Urea is typically used at a concentration of 8 M for protein denaturation or solubilization. A final concentration of 5 M urea is commonly used in molecular biology for sequencing gels. To prevent carbamylation, do not heat urea containing buffers above 37 °C.


One gram dissolves in 1 mL water, 10 mL 95% ethanol (100 mg/mL), 1 mL boiling 95% ethanol, 20 mL absolute ethanol(50 mg/mL), 6 mL methanol(166 mg/mL), 2 mL glycerol(500 mg/mL); Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether. Solutions can be treated to remove isocyanic acid. Acidification (100 mM HCl) can be used to drive the equilibrium to favor urea over the isocyanic acid, or a urea solution can be bulk deionised using an ion exchange resin.


Caution: Solutions of urea develop a significant concentration of reactive cyanate ions on standing. MP recommends making solutions fresh for each use.

Formula: NH₂CONH₂
MW: 60.06 g/mol
Melting Pt: 130…140 °C
MDL Number: MFCD00008022
CAS Number: 57-13-6
https://us.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg

Order Now

Specification Test Results

Synonyms Carbamide; Carbonyldiamine
Form White Powder
Storage Store at Room Temperature (15-30 °C).
Heavy Metals 10 ppm
pH 8.5 ± 1.0 (7M aq soln)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR