8-Anilino-1-naphthalenesulfonic acid, dark powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 8-Anilinonaphthalene-1-sulfonic acid, 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulfonic acid

0210563301 0210563305 0210563310 0210563325
IC10563301EA 46.34 USD
IC10563301 IC10563305 IC10563310 IC10563325
8-Anilino-1-naphthalenesulfonic acid, dark powder
8-Anilino-1-naphthalenesulphonic acid

8-Anilino-1-naphthalene Sulfonic Acid is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins.


ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants. It is a hydrophobic fluorescent probe for undenatured proteins on polyacrylamide gels.


Soluble in DMF (50mg/mL). Soluble in 1N NaOH, methanol (50 mg/mL - clear brownish green solution), chloroform (30 mg/mL in a 1:1 mixture produces a clear olive green solution); slightly water soluble.


Caution: Research Use Only (RUO).

Formula: C₁₆H₁₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00003998
CAS Number: 82-76-8

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR