Urea, white prills, Reagent Grade

Supplier: 0000012207

02103209.5 0210320905 0210320901
IC103209.5EA 48.24 USD
IC103209.5 IC10320905 76102-586
Urea, white prills, Reagent Grade
Urea

Urea is a prinicipal protein metabolite end product of nitrogen metabolism in most mammals, formed by the enzymatic reactions of the Kreb's cycle and the major product for the removal of free ammonia (NH4+) in vivo.


  • Vapor Pressure: 0.000016 hPa at 25 °C(Lit.)


Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels.
Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses.
Urea is used in cell or tissue culture media to increase the osmolality.
Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.


One gram dissolves in 1 mL water, 10 mL 95% ethanol (100 mg/mL), 1 mL boiling 95% ethanol, 20 mL absolute ethanol(50 mg/mL), 6 mL methanol(166 mg/mL), 2 mL glycerol(500 mg/mL); Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.


Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels. Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses. Urea is used in cell or tissue culture media to increase the osmolality. Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.

Formula: NH₂CONH₂
Boiling Pt: 135 °C (275 °F) (dec., lit.)
Melting Pt: 130…140 °C
Density: 1.335 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=995bef5c1f3be4c03122e42ad5ef32a51b8aa86b&vid=995bef5c1f3be4c03122e42ad5ef32a51b8aa86b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
OD260 nm (7 M aq soln) ≤0.2
OD280 (7M aq soln) ≤0.2
pH (7 M aq soln) 7.5-9.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR