L(-)-Tyrosine, off-white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
L-tyrosine is one of the twenty major amino acids. It is considered to be an essential amino acid. It is one of three amino acids with a bulky, uncharged, aromatic side group, a phenol group.
L-Tyrosine has been shown to assist in the production of melanin, the pigment found in hair and skin. Tyrosine is also used by the thyroid and adrenal glands to manufacture natural hormones, which regulate the body's ability to produce energy. Tyrosine is a precursor to dopamine, and therefore, plays a role in neurotranmission signaling and the regulation of depression. It also serves to release the appetite suppressing hormone, CCK, and assists in combatting allergic reactions.
Solubility in water ~45mg/100 mL. Soluble in alkaline and acidic aqueous solutions.
L-tyrosine is one of the twenty major amino acids. It is considered to be an essential amino acid. It is one of three amino acids with a bulky, uncharged, aromatic side group, a phenol group.
Caution: Research Use Only (RUO).
Formula:
C₉H₁₁NO₃ Melting Pt: 343 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002606 CAS Number: 60-18-4 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
UV/VIS: λ max (wate) | 270 - 275 nm |
Optical Rotation (c=4, 1N HCl) | -9.0° to -12.0° |
Loss on Drying | ≤0.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...